missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetyldiphenyl sulfone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.
Notes
Incompatible with strong oxidizing agents.
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 138°C to 140°C |
Quantity | 1 g |
Formula Weight | 260.31 |
Percent Purity | 98% |
Chemical Name or Material | 4-Acetyldiphenyl sulfone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'