Få mere at vide
4-Acetylbutyric Acid-d5 (Major), TRC
Artikelnummer. 30314582
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30314582 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314582 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168442100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Acetylbutyric Acid-d5 (Major)
CAS 1246817-13-9
Molekylær formel C6H5D5O3
Mængde 100 mg
Synonym 5-Oxo-hexanoic Acid-d5, 5-Ketocaproic Acid-d5, 5-Ketohexanoic Acid-d5, 5-Oxocaproic Acid-d5, 5-Oxohexanoic Acid-d5, NSC 5281-d5
InChI formel InChI=1S/C6H10O3/c1-5(7)3-2-4-6(8)9/h2-4H2,1H3,(H,8,9)/i1D3,3D2
SMIL [2H]C([2H])([2H])C(=O)C([2H])([2H])CCC(=O)O
IUPAC navn 4,4,6,6,6-pentadeuterio-5-oxohexanoic acid
Molekylvægt (g/mol) 135.17
Formel vægt 135.0944