Learn More
4-Acetylbenzoic Acid, Mikromol™
Product Code. 30050458
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050458 25 mg 25mg
30051948 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050458 Supplier Mikromol Supplier No. MM0002.080025
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Acetylbenzoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Acetylbenzoic Acid
CAS 586-89-0
Analyte or Component Names 4-Acetylbenzoic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H8 O3
Certifications/Compliance ISO 9001
Synonym 4-Acetylbenzoic Acid
InChI Formula InChI=1S/C9H8O3/c1-6(10)7-2-4-8(5-3-7)9(11)12/h2-5H,1H3,(H,11,12)
SMILES CC(=O)c1ccc(cc1)C(=O)O
IUPAC Name 4-acetylbenzoic acid
Molecular Weight (g/mol) 164.16
Quantity 25 mg
Formula Weight 164.0473 g/mol