Få mere at vide
4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Mikromol™
Artikelnummer. 30458556
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458556 4 x 25 mg
30050248 100 mg
30052216 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458556 Leverandør Mikromol Leverandørnr. MM0073.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
CAS 5608-13-9
Analyt- eller komponentnavne 4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
Forsendelsestilstand Room Temperature
Molekylær formel C16 H24 Cl N3 O3
Certificeringer/compliance ISO 17025
Synonym Metoclopramide Imp. A (EP), Metoclopramide USP Related Compound A, Metoclopramide USP RC A, 4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Metoclopramide Related Compound A, Metoclopramide Hydrochloride Monohydrate Impurity A, Metoclopramide Impurity A
InChI formel InChI=1S/C16H24ClN3O3/c1-5-20(6-2)8-7-18-16(22)12-9-13(17)14(19-11(3)21)10-15(12)23-4/h9-10H,5-8H2,1-4H3,(H,18,22)(H,19,21)
Løsningstype Neat
SMIL CCN(CC)CCNC(=O)c1cc(Cl)c(NC(=O)C)cc1OC
IUPAC navn 4-acetamido-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
Molekylvægt (g/mol) 341.83
Mængde 4 x 25 mg
Formel vægt 341.1506 g/mol