missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-4'-nitrodiphenyl ether, 97%, Thermo Scientific™

Catalog Number 11488997
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
75919-92-5
C14H11NO4
257.245
MFCD00052956
IRWXEFXORSDYQA-UHFFFAOYSA-N
4-acetyl-4'-nitrodiphenyl ether, 1-4-4-nitrophenoxy phenyl ethanone, 1-4-4-nitrophenoxy phenyl ethan-1-one, 1-acetyl-4-4-nitrophenoxy benzene, maybridge1_003157, cbmicro_014450, cambridge id 5228160, 4-acetyl-4-nitrodiphenylether, 4-4-acetylphenoxy nitrobenzene
238604
1-[4-(4-nitrophenoxy)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)OC2=CC=C(C=C2)[N+](=O)[O-]
This item is not returnable. View return policy
75919-92-5
C14H11NO4
257.245
MFCD00052956
IRWXEFXORSDYQA-UHFFFAOYSA-N
4-acetyl-4'-nitrodiphenyl ether, 1-4-4-nitrophenoxy phenyl ethanone, 1-4-4-nitrophenoxy phenyl ethan-1-one, 1-acetyl-4-4-nitrophenoxy benzene, maybridge1_003157, cbmicro_014450, cambridge id 5228160, 4-acetyl-4-nitrodiphenylether, 4-4-acetylphenoxy nitrobenzene
238604
1-[4-(4-nitrophenoxy)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)OC2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 79°C to 81°C
Quantity 1 g
Beilstein 2135124
Formula Weight 257.25
Percent Purity 97%
Chemical Name or Material 4-Acetyl-4'-nitrodiphenyl ether
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'