missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15455176
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1022154-80-8
C14H17BF2O3
282.094
MFCD22419259
NDJMQXWRZUTNSA-UHFFFAOYSA-N
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
68668352
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F
This item is not returnable. View return policy
1022154-80-8
C14H17BF2O3
282.094
MFCD22419259
NDJMQXWRZUTNSA-UHFFFAOYSA-N
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
68668352
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 72°C to 74°C
Quantity 250 mg
Formula Weight 282.09
Percent Purity 96%
Chemical Name or Material 4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'