missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 96%
Brand: Alfa Aesar™ H59660.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1022154-80-8 | |
282.094 | |
NDJMQXWRZUTNSA-UHFFFAOYSA-N | |
68668352 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F |
C14H17BF2O3 | |
MFCD22419259 | |
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone | |
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone |
Specifications
4-Acetyl-2,5-difluorobenzeneboronic acid pinacol ester | |
1g | |
C14H17BF2O3 | |
4-acetyl-2,5-difluorobenzeneboronic acid pinacol ester, 1-2,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2F)C(=O)C)F | |
282.094 | |
282.09 |
72°C to 74°C | |
1022154-80-8 | |
MFCD22419259 | |
NDJMQXWRZUTNSA-UHFFFAOYSA-N | |
1-[2,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone | |
68668352 | |
96% |