missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxy-2-azetidinone, 97%, Thermo Scientific Chemicals

Catalog Number p-7077627
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
28562-53-0
C5H7NO3
129.12
MFCD00010593
OEYMQQDJCUHKQS-UHFFFAOYNA-N
4-acetoxy-2-azetidinone, 2-azetidinone, 4-acetyloxy, 4-acetoxyazetidinone, 4-oxoazetidin-2-yl acetate, 2-oxoazetidinium 4-acetate, 2-azetidinone, 4-acetyloxy-, 4r, 4-acetoxy-azetidinone, acmc-20bsq9, 4-acetoxyazetidin-2-one, 4-acetoxyazetidine-2-one
119981
(4-oxoazetidin-2-yl) acetate
CC(=O)OC1CC(=O)N1
This item is not returnable. View return policy
28562-53-0
C5H7NO3
129.12
MFCD00010593
OEYMQQDJCUHKQS-UHFFFAOYNA-N
4-acetoxy-2-azetidinone, 2-azetidinone, 4-acetyloxy, 4-acetoxyazetidinone, 4-oxoazetidin-2-yl acetate, 2-oxoazetidinium 4-acetate, 2-azetidinone, 4-acetyloxy-, 4r, 4-acetoxy-azetidinone, acmc-20bsq9, 4-acetoxyazetidin-2-one, 4-acetoxyazetidine-2-one
119981
(4-oxoazetidin-2-yl) acetate
CC(=O)OC1CC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35.0°C to 38.0°C
Elemental Analysis 44.65 to 48.37% (C) 10.41 to 11.28% (N)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.12
Percent Purity 97%
Chemical Name or Material 4-Acetoxy-2-azetidinone
compliance-icons compliance-icons
Product Identifier
  • 4-Acetoxy-2-azetidinone, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'