Få mere at vide
4-Acetaminophen-d3 Sulfate Potassium Salt, TRC
Artikelnummer. 30324201
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30324201 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324201 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1612321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Acetaminophen-d3 Sulfate Potassium Salt
CAS 1188263-45-7
Molekylær formel C8H5D3KNO5S
Mængde 1 mg
Synonym N-[4-(Sulfooxy)phenyl]acetamide-d3 Monopotassium Salt, 4-Hydroxyacetanilide-d3 Sulfate Potassium Salt, PCM-S-d3 Potassium Salt, Paracetamol-d3 Sulfate Potassium Salt, Potassium N-Acetyl-p-aminophenyl-d3 Sulfate
InChI formel InChI=1S/C8H9NO5S.K/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13;/h2-5H,1H3,(H,9,10)(H,11,12,13);/q;+1/p-1/i1D3;
SMIL O=C(C([2H])([2H])[2H])NC1=CC=C(OS(=O)(O[K])=O)C=C1
IUPAC navn potassium;[4-[(2,2,2-trideuterioacetyl)amino]phenyl] sulfate
Molekylvægt (g/mol) 272.33