Få mere at vide
4-Acetamidocyclohexanol-d10, TRC
Artikelnummer. 30325358
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30325358 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325358 Leverandør Toronto Research Chemicals Leverandørnr. TRCA15843210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Acetamidocyclohexanol-d10
CAS 1241045-79-3
Molekylær formel C8H5D10NO2
Mængde 10 mg
Synonym N-(4-Hydroxycyclohexyl)acetamide-d10, NSC 130812-d10
InChI formel InChI=1S/C8H15NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h7-8,11H,2-5H2,1H3,(H,9,10)/i2D2,3D2,4D2,5D2,7D,8D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])(NC(=O)C)C([2H])([2H])C([2H])([2H])C1([2H])O
IUPAC navn N-(1,2,2,3,3,4,5,5,6,6-decadeuterio-4-hydroxycyclohexyl)acetamide
Molekylvægt (g/mol) 167.27
Formel vægt 167.173