missing translation for 'onlineSavingsMsg'
Få mere at vide

4-Acetamidobenzene-d4-sulfonyl Chloride, TRC

Artikelnummer. 30310335
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310335 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310335 Leverandør Toronto Research Chemicals Leverandørnr. TRCA170951100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-Acetamidobenzene-d4-sulfonyl Chloride
CAS 77435-44-0
Molekylær formel C8 D4 H4 Cl N O3 S
Mængde 100 mg
Synonym 4-(Acetylamino)benzenesulfonyl-d4 Chloride, 4-(Acetylamino)benzene-2,3,5,6-sulfonyl-d4 Chloride, NSC 127860-d4, ASC-d4, 4-(Acetylamino)phenylsulfonyl-d4 Chloride, p-Acetamidophenylsulfonyl-d4 Chloride, N-Acetylsulfanilyl-d4 Chloride, Dagenan-d4 Chloride, N-(4-Chlorosulfonylphenyl)acetamide-d4
InChI formel InChI=1S/C8H8ClNO3S/c1-6(11)10-7-2-4-8(5-3-7)14(9,12)13/h2-5H,1H3,(H,10,11)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Cl
IUPAC navn 4-acetamido-2,3,5,6-tetradeuteriobenzenesulfonyl chloride
Molekylvægt (g/mol) 237.7
Formel vægt 237.0164
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.