Få mere at vide
4-Acetamidobenzene-d4-sulfonyl Chloride, TRC
Artikelnummer. 30321960
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30321960 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321960 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17095150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Acetamidobenzene-d4-sulfonyl Chloride
CAS 77435-44-0
Molekylær formel C8 D4 H4 Cl N O3 S
Mængde 50 mg
Synonym 4-(Acetylamino)benzenesulfonyl-d4 Chloride, 4-(Acetylamino)benzene-2,3,5,6-sulfonyl-d4 Chloride, NSC 127860-d4, ASC-d4, 4-(Acetylamino)phenylsulfonyl-d4 Chloride, p-Acetamidophenylsulfonyl-d4 Chloride, N-Acetylsulfanilyl-d4 Chloride, Dagenan-d4 Chloride, N-(4-Chlorosulfonylphenyl)acetamide-d4
InChI formel InChI=1S/C8H8ClNO3S/c1-6(11)10-7-2-4-8(5-3-7)14(9,12)13/h2-5H,1H3,(H,10,11)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)C)S(=O)(=O)Cl
IUPAC navn 4-acetamido-2,3,5,6-tetradeuteriobenzenesulfonyl chloride
Molekylvægt (g/mol) 237.7
Formel vægt 237.0164