Få mere at vide
4-Acetamido Antipyrine-d3, TRC
Artikelnummer. 30320486
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30320486 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320486 Leverandør Toronto Research Chemicals Leverandørnr. TRCA14150250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Acetylaminoantipyrine D3
CAS 342821-66-3
Renhedsgrad noter HPLC
Molekylær formel C13 2H3 H12 N3 O2
Mængde 50 mg
Synonym 4-Acetylaminoantipyrine (acetyl-D3)
InChI formel InChI=1S/C13H15N3O2/c1-9-12(14-10(2)17)13(18)16(15(9)3)11-7-5-4-6-8-11/h4-8H,1-3H3,(H,14,17)/i2D3
SMIL [2H]C([2H])([2H])C(=O)NC1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC navn 2,2,2-trideuterio-N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide
Molekylvægt (g/mol) 248.3
Formel vægt 248.1353
Procent renhed >95