Få mere at vide
4,7-seco-Tebipenemoic Acid Methyl Ester Pivoxil, TRC
Artikelnummer. 30419048
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419048 2.5 mg
30414522 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30419048 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0128602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391053-29-4
Molekylær formel C23 H35 N3 O7 S2
Molekylvægt (g/mol) 529.67
IUPAC navn 2,2-dimethylpropanoyloxymethyl (2S,3R)-4-[1-(4,5-dihydro-1,3-thiazol-2-yl)azetidin-3-yl]sulfanyl-2-[(2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-yl]-3-methyl-2,3-dihydro-1H-pyrrole-5-carboxylate
SMIL COC(=O)[C@H]([C@@H](C)O)[C@@H]1NC(=C(SC2CN(C2)C3=NCCS3)[C@@H]1C)C(=O)OCOC(=O)C(C)(C)C
Tekniske data
Kemisk navn eller materiale 4,7-seco-Tebipenemoic Acid Methyl Ester Pivoxil
Mængde 2.5 mg
InChI formel InChI=1S/C23H35N3O7S2/c1-12-16(15(13(2)27)19(28)31-6)25-17(20(29)32-11-33-21(30)23(3,4)5)18(12)35-14-9-26(10-14)22-24-7-8-34-22/h12-16,25,27H,7-11H2,1-6H3/t12-,13-,15-,16-/m1/s1
Formel vægt 529.192