Få mere at vide
4-(6-Nitro-3-pyridinyl)-piperazine-2,2,6,6-d4, TRC
Artikelnummer. 30483818
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30483818 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483818 Leverandør Toronto Research Chemicals Leverandørnr. TRCN51993725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(6-Nitro-3-pyridinyl)-piperazine-2,2,6,6-d4
CAS 2089333-08-2
Molekylær formel C9 H9 F3 N2 O2
Mængde 25 mg
Synonym 1-(6-Nitro-3-pyridinyl)piperazine-d4, 1-(6-Nitropyridin-3-yl)piperazine-d4
InChI formel InChI=1S/C9H9F3N2O2/c1-3-16-7(15)6-4-13-8(9(10,11)12)14-5(6)2/h4H,3H2,1-2H3
SMIL CCOC(=O)c1cnc(nc1C)C(F)(F)F
IUPAC navn ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
Molekylvægt (g/mol) 234.175
Formel vægt 234.062