Få mere at vide
4-(6-Nitro-3-pyridinyl)-1-piperazine-2,2,6,6-d4-carboxylic Acid 1,1-Dimethylethyl Ester, TRC
Artikelnummer. 30482053
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30482053 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482053 Leverandør Toronto Research Chemicals Leverandørnr. TRCN51994725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(6-Nitro-3-pyridinyl)-1-piperazine-2,2,6,6-d4-carboxylic Acid 1,1-Dimethylethyl Ester
CAS 2089333-15-1
Molekylær formel C14H16D4N4O4
Mængde 25 mg
Synonym 4-(6-Nitro-3-pyridinyl)-1-piperazinecarboxylic Acid-d4 1,1-Dimethylethyl Ester, 4-(6-Nitropyridin-3-yl)piperazine-1-carboxylic-d4 Acid t-Butyl Ester
InChI formel InChI=1S/C14H20N4O4/c1-14(2,3)22-13(19)17-8-6-16(7-9-17)11-4-5-12(15-10-11)18(20)21/h4-5,10H,6-9H2,1-3H3/i8D2,9D2
SMIL CC(C)(C)OC(N1C([2H])([2H])CN(C2=CN=C(N(=O)=O)C=C2)CC1([2H])[2H])=O
IUPAC navn tert-butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate-2,2,6,6-d4
Molekylvægt (g/mol) 312.36
Formel vægt 312.17