Få mere at vide
4,6-Dimethyl-8-tert-butylcoumarin, TRC
Artikelnummer. 30376538
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376538 1 g
30374070 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30376538 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4714351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17874-34-9
Molekylær formel C15 H18 O2
Molekylvægt (g/mol) 230.3
Synonym Coumarin, 8-tert-butyl-4,6-dimethyl- (8CI), 8-(1,1-Dimethylethyl)-4,6-dimethyl-2H-1-benzopyran-2-one, 4,6-Dimethyl-8-tert-butylcoumarin, 8-(tert-Butyl)-4,6-dimethyl-2H-chromen-2-one
IUPAC navn 8-tert-butyl-4,6-dimethylchromen-2-one
SMIL CC1=CC(=O)Oc2c1cc(C)cc2C(C)(C)C
Tekniske data
Kemisk navn eller materiale 4,6-Dimethyl-8-tert-butylcoumarin
Mængde 1 g
InChI formel InChI=1S/C15H18O2/c1-9-6-11-10(2)8-13(16)17-14(11)12(7-9)15(3,4)5/h6-8H,1-5H3
Formel vægt 230.1307