Få mere at vide
4,6-Dimethyl-2(1H)-pyrimidinone Hydrochloride, TRC
Artikelnummer. 30372058
Skift visning
Mængde:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372058 25 g
30370539 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30372058 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47908025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34289-60-6
Molekylær formel C6H9ClN2O
Molekylvægt (g/mol) 160.6
Synonym 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride, 4,6-Dimethyl-2(1H)-pyrimidinone Hydrochloride, 4,6-Dimethylpyrimidin-2-ol Hydrochloride
SMIL Cl.Cc1cc(C)nc(O)n1
Tekniske data
Kemisk navn eller materiale 4,6-Dimethyl-2(1H)-pyrimidinone Hydrochloride
Mængde 25 g
InChI formel InChI=1S/C6H8N2O.ClH/c1-4-3-5(2)8-6(9)7-4;/h3H,1-2H3,(H,7,8,9);1H
Formel vægt 160.0403