Få mere at vide
4,6-Dichloro-5-nitro-2-(propylthio)pyrimidine, TRC
Artikelnummer. 30007223
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007223 250 mg
30004102 100 mg
30004485 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007223 Leverandør Toronto Research Chemicals Leverandørnr. TRCD436750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 145783-14-8
Molekylær formel C7 H7 Cl2 N3 O2 S
Molekylvægt (g/mol) 268.12
Synonym 4,6-Dichloro-5-nitro-2-propylthiopyrimidine
IUPAC navn 4,6-dichloro-5-nitro-2-propylsulfanyl-pyrimidine
SMIL CCCSc1nc(Cl)c(c(Cl)n1)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 4,6-Dichloro-5-nitro-2-(propylthio)pyrimidine
Mængde 250 mg
InChI formel InChI=1S/C7H7Cl2N3O2S/c1-2-3-15-7-10-5(8)4(12(13)14)6(9)11-7/h2-3H2,1H3
Formel vægt 266.9636