Få mere at vide
4,6-Dichloro-1,3-dinitrobenzene-13C6, CDN
Artikelnummer. 30325999
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30325999 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325999 Leverandør CDN Leverandørnr. CDNC50320.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dichloro-4,6-dinitrobenzene-13C6
CAS 335081-12-4
Molekylær formel 13C6 H2 Cl2 N2 O4
Mængde 0.1 g
Synonym Benzene-13C6, 1,5-dichloro-2,4-dinitro- (9 CI), 1,5-Dichloro-2,4-dinitrobenzene-13C6, 1,3-Dichloro-4,6-dinitrobenzene-13C6, 2,4-Dichloro-1,5-dinitrobenzene-13C6, 2,6-Dichloro-3,5-dinitrobenzene-13C6, 4,6-Dichloro-1,3-dinitrobenzene-13C6, 4,6-Dinitro-1,3-dichlorobenzene-13C6
InChI formel InChI=1 S/C6H2Cl2N2O4/c7-3-1-4(8)6(10(13)14)2-5(3)9(11)12/h1-2 H/i1+1,2+1,3+1,4+1,5+1,6+1
SMIL [O-][N+](=O)[13 c]1[13 cH][13 c]([13 c](Cl)[13 cH][13 c]1 Cl)[N+](=O)[O-]
IUPAC navn 1,5-dichloro-2,4-dinitro(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene
Molekylvægt (g/mol) 242.953
Formel vægt 241.9593 g/mol
Procent renhed 99 atom % 13 C, min 98% Chemical Purity