Få mere at vide
(4,6-Diamino-1,3,5-triazin-2-yl)guanidine Nitrate, Mikromol™
Beskrivelse
(4,6-Diamino-1,3,5-triazin-2-yl)guanidine Nitrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (4,6-Diamino-1,3,5-triazin-2-yl)guanidine Nitrate |
| CAS | 3553-49-9 |
| Analyt- eller komponentnavne | (4,6-Diamino-1,3,5-triazin-2-yl)guanidine Nitrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C4 H8 N8 . H N O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Metformin Hydrochloride Imp. B (EP), Metformin Imp. B (EP), (4,6-Diamino-1,3,5-triazin-2-yl)guanidine Nitrate, Metformin Hydrochloride Imp. B (EP) as Nitrate, Metformin Hydrochloride Impurity B as Nitrate, Metformin Impurity B as Nitrate |
| InChI formel | InChI=1S/C4H8N8.HNO3/c5-1(6)9-4-11-2(7)10-3(8)12-4;2-1(3)4/h(H8,5,6,7,8,9,10,11,12);(H,2,3,4) |
| SMIL | NC(=N)Nc1nc(N)nc(N)n1.O[N+](=O)[O-] |
| IUPAC navn | 1-(4,6-diamino-1,3,5-triazin-2-yl)guanidine;nitric acid |
| Vis mere |