Få mere at vide
4-[[6-Chloro-2-[(4-cyanophenyl)amino]-4-pyrimidinyl]oxy]-3,5-dimethylbenzonitrile-d8, TRC
Artikelnummer. 30364096
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30364096 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30364096 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3808245MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-((6-Chloro-2-((4-cyanophenyl)amino)-4-pyrimidinyl)oxy)-3,5-dimethylbenzonitrile-d8
CAS 1142096-20-5
Molekylær formel C20 D8 H6 Cl N5 O
Mængde 5 mg
Synonym 4-[[6-Chloro-2-[(4-cyanophenyl)amino]-4-pyrimidinyl]oxy]-3,5-dimethyl-benzonitrile-d8
InChI formel InChI=1S/C20H14ClN5O/c1-12-7-15(11-23)8-13(2)19(12)27-18-9-17(21)25-20(26-18)24-16-5-3-14(10-22)4-6-16/h3-9H,1-2H3,(H,24,25,26)/i1D3,2D3,7D,8D
SMIL [2H]c1c(C#N)c([2H])c(c(Oc2cc(Cl)nc(Nc3ccc(cc3)C#N)n2)c1C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 4-[6-chloro-2-(4-cyanoanilino)pyrimidin-4-yl]oxy-2,6-dideuterio-3,5-bis(trideuteriomethyl)benzonitrile
Molekylvægt (g/mol) 383.86
Formel vægt 383.139