missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Diphenylimidazole, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.
Solubility
Soluble in methanol.
Notes
Incompatible with strong acids.
4,5-Diphenylimidazole is used as a precursor for azo dyes such as 2-(quinolylazo)-4,5-diphenylimidazole . It is also used to determine metal ions by spectrophotometrically. It acts as a chromogenic reagent.
Solubility
Soluble in methanol.
Notes
Incompatible with strong acids.
Specifications
Melting Point | 230°C to 234°C |
Quantity | 25 g |
Beilstein | 157587 |
Solubility Information | Soluble in methanol. |
Formula Weight | 220.28 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,5-Diphenylimidazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'