Få mere at vide
4,5-Diphenyl-2-oxazolepropanamide, TRC
Artikelnummer. 30375083
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30375083 500 mg
30369303 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30375083 Leverandør Toronto Research Chemicals Leverandørnr. TRCD491610500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 24248-49-5
Molekylær formel C18H16N2O2
Molekylvægt (g/mol) 292.33
Synonym 3-(4,5-Diphenyloxazol-2-yl)propanamide, beta-(4,5-Diphenyloxazol-2-yl)propionamide, Wy 22075
SMIL NC(=O)CCc1oc(c2ccccc2)c(n1)c3ccccc3
Tekniske data
Kemisk navn eller materiale 4,5-Diphenyl-2-oxazolepropanamide
Mængde 500 mg
InChI formel InChI=1S/C18H16N2O2/c19-15(21)11-12-16-20-17(13-7-3-1-4-8-13)18(22-16)14-9-5-2-6-10-14/h1-10H,11-12H2,(H2,19,21)
Formel vægt 292.1212