missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoroanthranilic acid, 97%, Thermo Scientific™

Catalog Number p-3771089
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
83506-93-8
C7H5F2NO2
173.12
MFCD00077509
DGOZIZVTANAGCA-UHFFFAOYSA-N
4,5-difluoroanthranilic acid, 2-amino-4,5-difluoro-benzoic acid, buttpark 49\07-41, benzoicacid, 2-amino-4,5-difluoro, benzoic acid, 2-amino-4,5-difluoro, pubchem4675, 2-amino-4,5-difluoro, 4,5-difluoroantranilic acid, ksc448a3l, 4,5-difluoro anthranilic acid
735965
2-amino-4,5-difluorobenzoic acid
NC1=CC(F)=C(F)C=C1C(O)=O
This item is not returnable. View return policy
83506-93-8
C7H5F2NO2
173.12
MFCD00077509
DGOZIZVTANAGCA-UHFFFAOYSA-N
4,5-difluoroanthranilic acid, 2-amino-4,5-difluoro-benzoic acid, buttpark 49\07-41, benzoicacid, 2-amino-4,5-difluoro, benzoic acid, 2-amino-4,5-difluoro, pubchem4675, 2-amino-4,5-difluoro, 4,5-difluoroantranilic acid, ksc448a3l, 4,5-difluoro anthranilic acid
735965
2-amino-4,5-difluorobenzoic acid
NC1=CC(F)=C(F)C=C1C(O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 181.0°C to 183.0°C
Color Colorless to Straw
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula F2C6H2(NH2)COOH
Quantity 1 g
Formula Weight 173.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 5-Difluoroanthranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Difluoroanthranilic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'