missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-methoxybenzylamine, 97%, Thermo Scientific™

Catalog Number 15493447
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886497-93-4
C8H9F2NO
173.16
MFCD06660190
LZSNRJCSFRPZSW-UHFFFAOYSA-N
4,5-difluoro-2-methoxybenzylamine, 1-4,5-difluoro-2-methoxyphenyl methanamine, 4,5-difluoro-2-methoxy-benzylamine, 4,5-difluoro-2-methoxyphenyl methanamine, 4,5-difluoro-2-methoxyphenyl methylamine
17750766
(4,5-difluoro-2-methoxyphenyl)methanamine
COC1=CC(F)=C(F)C=C1CN
This item is not returnable. View return policy
886497-93-4
C8H9F2NO
173.16
MFCD06660190
LZSNRJCSFRPZSW-UHFFFAOYSA-N
4,5-difluoro-2-methoxybenzylamine, 1-4,5-difluoro-2-methoxyphenyl methanamine, 4,5-difluoro-2-methoxy-benzylamine, 4,5-difluoro-2-methoxyphenyl methanamine, 4,5-difluoro-2-methoxyphenyl methylamine
17750766
(4,5-difluoro-2-methoxyphenyl)methanamine
COC1=CC(F)=C(F)C=C1CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.505
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 4,5-Difluoro-2-methoxybenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'