missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-2-fluorotoluene, 98%, Thermo Scientific™

Catalog Number p-7054889
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
202982-77-2
C7H5Br2F
267.923
MFCD00143234
GJHJZTCWLKJMIG-UHFFFAOYSA-N
4,5-dibromo-2-fluorotoluene, 2-fluoro-4,5-dibromotoluene, pubchem10411, benzene,1,2-dibromo-4-fluoro-5-methyl, 1,2-bis bromanyl-4-fluoranyl-5-methyl-benzene
2773986
1,2-dibromo-4-fluoro-5-methylbenzene
CC1=CC(=C(C=C1F)Br)Br
This item is not returnable. View return policy
202982-77-2
C7H5Br2F
267.923
MFCD00143234
GJHJZTCWLKJMIG-UHFFFAOYSA-N
4,5-dibromo-2-fluorotoluene, 2-fluoro-4,5-dibromotoluene, pubchem10411, benzene,1,2-dibromo-4-fluoro-5-methyl, 1,2-bis bromanyl-4-fluoranyl-5-methyl-benzene
2773986
1,2-dibromo-4-fluoro-5-methylbenzene
CC1=CC(=C(C=C1F)Br)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 267.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4,5-Dibromo-2-fluorotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'