Få mere at vide
4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, TRC
Artikelnummer. 30137042
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30137042 2.5 g 2.5g
17983555 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137042 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3749252.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346598-11-5
Molekylær formel C13 H14 Cl N3 O2
Molekylvægt (g/mol) 279.72
Synonym 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, Domperidone Imp. B (EP), 1-Piperidinecarboxaldehyde, 4-(5-chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)- (ACI), 4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinecarboxaldehyde (ACI)
IUPAC navn 4-(5-chloro-2-oxo-3H-benzimidazol-1-yl)piperidine-1-carbaldehyde
SMIL Clc1ccc2N(C3CCN(CC3)C=O)C(=O)Nc2c1
Tekniske data
Kemisk navn eller materiale 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
Mængde 2.5 g
InChI formel InChI=1S/C13H14ClN3O2/c14-9-1-2-12-11(7-9)15-13(19)17(12)10-3-5-16(8-18)6-4-10/h1-2,7-8,10H,3-6H2,(H,15,19)
Formel vægt 279.0775