Få mere at vide
4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, Mikromol™
Artikelnummer. 17901026
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17901026 25 mg
17960886 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17901026 Leverandør Mikromol Leverandørnr. MM0107.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
CAS 1346598-11-5
Analyt- eller komponentnavne 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine
Forsendelsestilstand Room Temperature
Molekylær formel C13 H14 Cl N3 O2
Certificeringer/compliance ISO 9001
Synonym Domperidone Imp. B (EP), 4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-formylpiperidine, Domperidone Impurity B
InChI formel InChI=1S/C22H28N2O2.ClH/c1-2-22(25)23(20-11-7-4-8-12-20)21-14-17-24(26,18-15-21)16-13-19-9-5-3-6-10-19;/h3-12,21H,2,13-18H2,1H3;1H
SMIL Clc1ccc2N(C3CCN(CC3)C=O)C(=O)Nc2c1
IUPAC navn 4-(5-chloro-2-oxo-3H-benzimidazol-1-yl)piperidine-1-carbaldehyde
Molekylvægt (g/mol) 279.72
Mængde 25 mg
Formel vægt 279.0775 g/mol