missing translation for 'onlineSavingsMsg'
Få mere at vide

4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile, Mikromol™

Artikelnummer. 30052271
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052271 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30052271 Leverandør Mikromol Leverandørnr. MM3297.070025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile
CAS 236750-65-5
Analyt- eller komponentnavne 4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile
Forsendelsestilstand Room Temperature
Molekylær formel C13 H16 N2 O6
Certificeringer/compliance ISO 9001
Synonym Benzonitrile, 4,5-bis(2-methoxyethoxy)-2-nitro-, 4,5-Bis(2-methoxyethoxy)-2-nitrobenzonitrile, 2-Nitro-4,5-(2-methoxyethoxy)benzonitrile, 4,5-Di(2-methoxyethoxy)-2-nitrobenzonitrile
InChI formel InChI=1S/C13H16N2O6/c1-18-3-5-20-12-7-10(9-14)11(15(16)17)8-13(12)21-6-4-19-2/h7-8H,3-6H2,1-2H3
SMIL COCCOc1cc(C#N)c(cc1OCCOC)[N+](=O)[O-]
IUPAC navn 4,5-bis(2-methoxyethoxy)-2-nitrobenzonitrile
Molekylvægt (g/mol) 296.2759
Mængde 25 mg
Formel vægt 296.1008 g/mol
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.