Få mere at vide
4,5,7-Trichloroquinoline, TRC
Artikelnummer. 30454918
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454918 1 g
30452041 500 mg
30451856 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30454918 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7747201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23834-01-7
Molekylær formel C9H4Cl3N
Molekylvægt (g/mol) 232.49
Synonym 4,5,7-Trichloro-quinoline
IUPAC navn 4,5,7-trichloroquinoline
SMIL ClC1=CC=NC2=CC(Cl)=CC(Cl)=C21
Tekniske data
Kemisk navn eller materiale 4,5,7-Trichloroquinoline
Mængde 1 g
InChI formel InChI=1S/C9H4Cl3N/c10-5-3-7(12)9-6(11)1-2-13-8(9)4-5/h1-4H
Formel vægt 230.94