missing translation for 'onlineSavingsMsg'
Learn More

4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 15424356
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
19156-54-8
C9H10O2S
182.237
MFCD00652575
TUZZQEHPGHKGRJ-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid
767738
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid
C1CCC2=C(C1)C(=CS2)C(=O)O
This item is not returnable. View return policy
19156-54-8
C9H10O2S
182.237
MFCD00652575
TUZZQEHPGHKGRJ-UHFFFAOYSA-N
4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydrobenzo b thiophene-3-carboxylicacid, benzo b thiophene-3-carboxylic acid, 4,5,6,7-tetrahydro, benzo b thiophene-3-carboxylicacid, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic a, 4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylicacid
767738
4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid
C1CCC2=C(C1)C(=CS2)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 1367376
Formula Weight 182.24
Percent Purity 97%
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'