Få mere at vide
4,5,6,7-Tetrabromobenzimidazole, TRC
Artikelnummer. 30410734
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410734 250 mg
30415099 100 mg
30412615 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30410734 Leverandør Toronto Research Chemicals Leverandørnr. TRCT291150250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 577779-57-8
Molekylær formel C7 H2 Br4 N2
Molekylvægt (g/mol) 433.72
IUPAC navn 4,5,6,7-tetrabromo-1H-benzimidazole
SMIL Brc1c(Br)c(Br)c2[nH]cnc2c1Br
Tekniske data
Kemisk navn eller materiale 4,5,6,7-Tetrabromobenzimidazole
Mængde 250 mg
InChI formel InChI=1S/C7H2Br4N2/c8-2-3(9)5(11)7-6(4(2)10)12-1-13-7/h1H,(H,12,13)
Formel vægt 429.6951