missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number 15428495
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1227955-11-4
C12H7ClF3NO3S
337.70
MFCD11109522
UXJUIRATKKZXFB-UHFFFAOYSA-N
4-4-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 2-4-chlorosulphonyl phenoxy-4-trifluoromethyl pyridine, 4-4-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, 4-4-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-4-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
51342061
4-[4-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
FC(F)(F)C1=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=NC=C1
This item is not returnable. View return policy
1227955-11-4
C12H7ClF3NO3S
337.70
MFCD11109522
UXJUIRATKKZXFB-UHFFFAOYSA-N
4-4-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 2-4-chlorosulphonyl phenoxy-4-trifluoromethyl pyridine, 4-4-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, 4-4-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-4-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride
51342061
4-[4-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
FC(F)(F)C1=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 77°C to 79°C
Quantity 250 mg
UN Number UN3261
Formula Weight 337.71
Percent Purity 97%
Chemical Name or Material 4-(4-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'