Få mere at vide
4,4'-Thiobis(2-tert-butyl-5-methylphenol), TRC
Tekniske data
Specifications
| Kemisk navn eller materiale | 4,4'-Thiobis(2-tert-butyl-5-methylphenol) |
| CAS | 96-69-5 |
| Molekylær formel | C22 H30 O2 S |
| Mængde | 25 g |
| Synonym | m-Cresol, 4,4'-thiobis[6-tert-butyl- (8CI), 4,4'-Thiobis[2-(1,1-dimethylethyl)-5-methylphenol], 1,1'-Thiobis(2-methyl-4-hydroxy-5-tert-butyl)benzene, 2,2'-Di-tert-butyl-5,5'-dimethyl-4,4'-thiodiphenol, 2-tert-Butyl-4-(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl-5-methylphenol, 4,4'-Bis(6-tert-butyl-m-cresol) sulfide, 4,4'-Thiobis(3-methyl-6-tert-butylphenol), 4,4'-Thiobis(6-t-butyl-m-cresol), 4,4'-Thiobis(6-tert-butyl-m-cresol), 4,4'-Thiobis[2-tert-butyl-5-methylphenol], 4,4'-Thiobis[3-methyl-6-tert-butylphenol], 4,4'-Thiobis[6-tert-butyl-3-methylphenol], 4,4'-Thiobis[6-tert-butyl-m-cresol], AO 1081, AO 300, AO 4, AO-TBM 6, Antage Crystal, Antigene WX-R, Antioxidant 300, Antioxidant B 102, Antioxidant BPS, Antioxidant TMB 6, Bis(2-methyl-4-hydroxy-5-tert-butylphenyl) sulfide, Bis(2-methyl-5-tert-butyl-4-hydroxyphenyl) sulfide, Bis(3-tert-butyl-4-hydroxy-6-methylphenyl) sulfide, Bis(4-hydroxy-5-tert-butyl-2-methylphenyl) sulfide, Bis(5-tert-butyl-4-hydroxy-2-methylphenyl) sulfide, Chinox 300, Disperse MB 61, Durad AX 16, Irganox 300, Irganox 415, K 300, Keminox 236T, Lowinox 44S36, Lowinox Rosin 6, Lowinox TBM 6, Lowinox TBM 6P, N 300, NSC 35388, Nocrac 300, Nonflex BPS, Nonflex BPS-R, Santonox, Santonox BM, Santonox R, Santonox TBMC, Santowhite Crystals, Seenox BCS, Sumilizer WX, Sumilizer WX-R, Sumilizer WX-RC, TBM 6, Thioalkofen BM, Thioalkofen BM 4, Thioalkofen BMCh, Thioalkofen MBCh, Ultranox 236, Yoshinox S, Yoshinox SR, ZBX 1R, 4,4'-Thiobis(2-tert-butyl-5-methylphenol) |
| InChI formel | InChI=1S/C22H30O2S/c1-13-9-17(23)15(21(3,4)5)11-19(13)25-20-12-16(22(6,7)8)18(24)10-14(20)2/h9-12,23-24H,1-8H3 |
| SMIL | Cc1cc(O)c(cc1Sc2cc(c(O)cc2C)C(C)(C)C)C(C)(C)C |
| IUPAC navn | 2-tert-butyl-4-(5-tert-butyl-4-hydroxy-2-methylphenyl)sulfanyl-5-methylphenol |
| Molekylvægt (g/mol) | 358.54 |
| Formel vægt | 358.1967 |