Få mere at vide
4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) Tetrahydrochloride, TRC
Artikelnummer. 30427740
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427740 100 mg
30422626 50 mg
30412113 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427740 Leverandør Toronto Research Chemicals Leverandørnr. TRCP835305100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74918-21-1
Molekylær formel C15H20N4O2 . 4(HCl)
Molekylvægt (g/mol) 434.2
IUPAC navn 4-[3-(2,4-diaminophenoxy)propoxy]benzene-1,3-diamine;tetrahydrochloride
SMIL Cl.Cl.Cl.Cl.Nc1ccc(OCCCOc2ccc(N)cc2N)c(N)c1
Tekniske data
Kemisk navn eller materiale 4,4'-(Propane-1,3-diylbis(oxy))bis(benzene-1,3-diamine) Tetrahydrochloride
Mængde 100 mg
InChI formel InChI=1S/C15H20N4O2.4ClH/c16-10-2-4-14(12(18)8-10)20-6-1-7-21-15-5-3-11(17)9-13(15)19;;;;/h2-5,8-9H,1,6-7,16-19H2;4*1H
Formel vægt 432.0653