Få mere at vide
4-(4-Phenylbutoxy)benzoic Acid, TRC
Artikelnummer. 30240705
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240705 50 mg
30240823 250 mg
30240877 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240705 Leverandør Toronto Research Chemicals Leverandørnr. TRCP39925050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 30131-16-9
Molekylær formel C17H18O3
Molekylvægt (g/mol) 270.32
IUPAC navn 4-(4-phenylbutoxy)benzoic acid
SMIL OC(=O)c1ccc(OCCCCc2ccccc2)cc1
Tekniske data
Kemisk navn eller materiale 4-(4-Phenylbutoxy)benzoic Acid
Mængde 50 mg
InChI formel InChI=1S/C17H18O3/c18-17(19)15-9-11-16(12-10-15)20-13-5-4-8-14-6-2-1-3-7-14/h1-3,6-7,9-12H,4-5,8,13H2,(H,18,19)
Formel vægt 270.1256