missing translation for 'onlineSavingsMsg'
Få mere at vide

4,4'-oxydibenzensulfonylchlorid, TRC

Product Code. 30428393
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30428393 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428393 Supplier Toronto Research Chemicals Supplier No. TRCO8706301G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4,4'-Oxydibenzenesulfonyl Chloride
CAS 121-63-1
Molekylær formel C12H8Cl2O5S2
Mængde 1 g
Synonym 4,4'-Oxybis[benzenesulfonyl Chloride], 4,4'-Bis(chlorosulfonyl)biphenyl Ether, 4,4'-Bis(chlorosulfonyl)diphenyl Ether, 4,4'-Oxybis(benzenesulfonyl Chloride), 4,4'-Oxydibenzenesulfonyl Chloride, Bis(4-Chlorosulfonylphenyl) Ether, Diphenyl Ether 4,4’-Disulfonyl Chloride, NSC 212, Oxybis(4-benzenesulfonyl Chloride), Oxybis(benzenesulfonyl Chloride), Phenoxybenzene-4,4'-disulfonyl Chloride, p,p'-Oxybis(benzenesulfonyl Chloride), 4,4’-Oxybisbenzenesulfonyl Chloride
InChI formel InChI=1S/C12H8Cl2O5S2/c13-20(15,16)11-5-1-9(2-6-11)19-10-3-7-12(8-4-10)21(14,17)18/h1-8H
SMIL ClS(=O)(=O)c1ccc(Oc2ccc(cc2)S(=O)(=O)Cl)cc1
IUPAC navn 4-(4-chlorosulfonylphenoxy)benzensulfonylchlorid
Molekylvægt (g/mol) 367.22
Formel vægt 365.919
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.