Få mere at vide
4-(4-Nitrophenyl)-3-morpholinone-d4, TRC
Artikelnummer. 30477032
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30477032 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477032 Leverandør Toronto Research Chemicals Leverandørnr. TRCN504427100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Nitrophenyl)-3-morpholinone-d4
CAS 1330180-47-6
Molekylær formel C10H6D4N2O4
Mængde 100 mg
Synonym 4-(3-Oxo-4-morpholinyl)nitrobenzene-d4, 4-(4-Nitrophenyl)-3-oxomorpholine-d4, 4-(4-Nitrophenyl)morpholin-3-one-d4
InChI formel InChI=1S/C10H10N2O4/c13-10-7-16-6-5-11(10)8-1-3-9(4-2-8)12(14)15/h1-4H,5-7H2/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1N2CCOCC2=O)[N+](=O)[O-]
IUPAC navn 4-(2,3,5,6-tetradeuterio-4-nitrophenyl)morpholin-3-one
Molekylvægt (g/mol) 226.22
Formel vægt 226.0892