missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)benzylamine, ≥90%, Thermo Scientific™

Catalog Number p-2699857
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
216144-45-5
C12H19N3
205.305
MFCD03086117
MZFQGKRIWIKPBT-UHFFFAOYSA-N
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
2776493
[4-(4-methylpiperazin-1-yl)phenyl]methanamine
CN1CCN(CC1)C2=CC=C(C=C2)CN
This item is not returnable. View return policy
216144-45-5
C12H19N3
205.305
MFCD03086117
MZFQGKRIWIKPBT-UHFFFAOYSA-N
4-4-methylpiperazino benzylamine, 4-4-methylpiperazin-1-yl benzylamine, 4-4-methylpiperazin-1-yl phenyl methanamine, 1-4-4-methylpiperazin-1-yl phenyl methanamine, 4-4-methyl-piperazin-1-yl-benzylamine, 4-4-methylpiperazin-1-yl-benzylamine, benzenemethanamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenyl methylamine, pubchem9053
2776493
[4-(4-methylpiperazin-1-yl)phenyl]methanamine
CN1CCN(CC1)C2=CC=C(C=C2)CN
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.3
Percent Purity 90%
Chemical Name or Material 4-(4-Methylpiperazino)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'