missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N,N-dimethylaniline), 98+%, Thermo Scientific Chemicals

Catalog Number p-7024178
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
101-61-1
C17H22N2
254.377
MFCD00008317
JNRLEMMIVRBKJE-UHFFFAOYSA-N
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
7567
CHEBI:34370
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C
This item is not returnable. View return policy
101-61-1
C17H22N2
254.377
MFCD00008317
JNRLEMMIVRBKJE-UHFFFAOYSA-N
4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
7567
CHEBI:34370
4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C

4,4'-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 85°C to 88°C
Boiling Point 155°C to 157°C (0.1 mmHg)
Flash Point 178°C (352°F)
Quantity 50 g
UN Number UN3077
Beilstein 479956
Merck Index 14,6176
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 254.38
Percent Purity ≥98%
Chemical Name or Material 4,4'-Methylenebis(N,N-dimethylaniline)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'