missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone, 98%, Thermo Scientific Chemicals

Catalog Number p-7021652
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
5471-51-2
C10H12O2
164.204
MFCD00002394
NJGBTKGETPDVIK-UHFFFAOYSA-N
4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
21648
CHEBI:68656
4-(4-hydroxyphenyl)butan-2-one
CC(=O)CCC1=CC=C(C=C1)O
This item is not returnable. View return policy
5471-51-2
C10H12O2
164.204
MFCD00002394
NJGBTKGETPDVIK-UHFFFAOYSA-N
4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
21648
CHEBI:68656
4-(4-hydroxyphenyl)butan-2-one
CC(=O)CCC1=CC=C(C=C1)O

4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 80°C to 84°C
Quantity 25 g
Beilstein 776080
Solubility Information Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'