Få mere at vide
4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridin Hydrochlorid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine Hydrochloride |
| CAS | 1012886-75-7 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C12 H14 F N . Cl H |
| Mængde | 10 mg |
| Synonym | Pyridine, 4-(4-fluorophenyl)-1,2,3,6-tetrahydro-1-methyl-, hydrochloride (1:1), 4-(4-Fluorophenyl)-1-methyl-1,2,3,6-tetrahydropyridine hydrochloride, 4-(4-Fluorophenyl)-1,2,3,6-tetrahydro-1-methylpyridine hydrochloride, 1-Methyl-4-(4'-fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, Paroxetine Hydrochloride Hemihydrate Imp. G (EP) as Hydrochloride |
| InChI formel | InChI=1S/C12H14FN.ClH/c1-14-8-6-11(7-9-14)10-2-4-12(13)5-3-10;/h2-6H,7-9H2,1H3;1H |
| SMIL | Cl.CN1CCC(=CC1)c2ccc(F)cc2 |
| IUPAC navn | 4-(4-fluorophenyl)-1-methyl-3,6-dihydro-2H-pyridin; hydrochlorid |
| Molekylvægt (g/mol) | 227.71 |
| Vis mere |