Få mere at vide
4,4'-Dipyridyl-d8, CDN
Artikelnummer. 30321508
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321508 0.25 g -
30319623 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321508 Leverandør CDN Leverandørnr. CDND50730.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,4'-Bipyridine-d8
CAS 132125-39-4
Molekylær formel C10 2H8 N2
Mængde 0.25 g
Synonym 4,4'-Bipyridine-2,2',3,3',5,5',6,6'-d8, 4,4'-Bipyridine-d8, SERS 421
InChI formel InChI=1 S/C10H8N2/c1-5-11-6-2-9(1)10-3-7-12-8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1nc([2 H])c([2 H])c(c1[2 H])c2c([2 H])c([2 H])nc([2 H])c2[2 H]
IUPAC navn 2,3,5,6-tetradeuterio-4-(2,3,5,6-tetradeuteriopyridin-4-yl)pyridine
Molekylvægt (g/mol) 164.23
Formel vægt 164.119 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity