Få mere at vide
4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-[4-[(2-hydroxy-1,1-dimethyl)ethyl]phenyl]butyne, TRC
Artikelnummer. 30374967
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30374967 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374967 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4917872.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-[4-[(2-hydroxy-1,1-dimethyl)ethyl]phenyl]butyne
CAS 1189696-09-0
Molekylær formel C32H32D5NO2
Mængde 2.5 mg
InChI formel InChI=1S/C32H37NO2/c1-32(2,25-34)29-18-16-26(17-19-29)11-9-10-22-33-23-20-30(21-24-33)35-31(27-12-5-3-6-13-27)28-14-7-4-8-15-28/h3-8,12-19,30-31,34H,10,20-25H2,1-2H3/i3D,5D,6D,12D,13D
SMIL [2H]c1c([2H])c([2H])c(C(OC2CCN(CCC#Cc3ccc(cc3)C(C)(C)CO)CC2)c4ccccc4)c([2H])c1[2H]
IUPAC navn 2-methyl-2-[4-[4-[4-[(2,3,4,5,6-pentadeuteriophenyl)-phenylmethoxy]piperidin-1-yl]but-1-ynyl]phenyl]propan-1-ol
Molekylvægt (g/mol) 472.67
Formel vægt 472.3138