missing translation for 'onlineSavingsMsg'
Få mere at vide

4,4-Dimethyldihydrofuran-2,3-dione, TRC

Artikelnummer. 30371343
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30371343 500 mg
30369717 100 mg
30385256 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371343 Supplier Toronto Research Chemicals Supplier No. TRCD476943500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-04-4
Molekylær formel C6H8O3
Molekylvægt (g/mol) 128.13
Synonym Dexpanthenol Impurity 5
IUPAC navn 4,4-dimethyloxolane-2,3-dione
SMIL CC1(C)COC(=O)C1=O

Specifications

Kemisk navn eller materiale 4,4-Dimethyldihydrofuran-2,3-dione
Mængde 500 mg
InChI formel InChI=1S/C6H8O3/c1-6(2)3-9-5(8)4(6)7/h3H2,1-2H3
Formel vægt 128.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.