missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzil, 98%, Thermo Scientific Chemicals

Catalog Number p-7024242
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3457-48-5
C16H14O2
238.286
MFCD00008554
BCWCEHMHCDCJAD-UHFFFAOYSA-N
4,4'-dimethylbenzil, 1,2-di-p-tolylethane-1,2-dione, p-tolil, 1,2-bis 4-methylphenyl ethane-1,2-dione, ethanedione, bis 4-methylphenyl, ethandione, bis p-tolyl, bis 4-methylphenyl ethane-1,2-dione, 1,2-bis 4-methylphenyl-1,2-ethanedione, di-p-tolylethanedione, 4,4/'-dimethylbenzil
76996
1,2-bis(4-methylphenyl)ethane-1,2-dione
CC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)C
This item is not returnable. View return policy
3457-48-5
C16H14O2
238.286
MFCD00008554
BCWCEHMHCDCJAD-UHFFFAOYSA-N
4,4'-dimethylbenzil, 1,2-di-p-tolylethane-1,2-dione, p-tolil, 1,2-bis 4-methylphenyl ethane-1,2-dione, ethanedione, bis 4-methylphenyl, ethandione, bis p-tolyl, bis 4-methylphenyl ethane-1,2-dione, 1,2-bis 4-methylphenyl-1,2-ethanedione, di-p-tolylethanedione, 4,4/'-dimethylbenzil
76996
1,2-bis(4-methylphenyl)ethane-1,2-dione
CC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 104°C
Quantity 1 g
Formula Weight 238.29
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethylbenzil

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'