missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-1-pentene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Density | 0.6820g/mL |
Boiling Point | 70°C to 72°C |
Flash Point | -12°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH2CHCH2C(CH3)3 |
Quantity | 1 g |
Specific Gravity | 0.682 |
Show More |
Safety and Handling
Product Identifier
- 4,4-Dimethyl-1-pentene
- Danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H304-May be fatal if swallowed and enters airways.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'