Få mere at vide
4,4'-Dimethoxystilbene (~90%), TRC
Artikelnummer. 30378460
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378460 100 mg
30370926 1 g
30376244 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30378460 Leverandør Toronto Research Chemicals Leverandørnr. TRCD479470100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4705-34-4
Molekylær formel C16 H16 O2
Molekylvægt (g/mol) 240.3
Synonym 1,1'-(1,2-ethenediyl)bis(4-methoxybenzene), Bianisal, Bianisylidene, Photoanethole, p,p'-Dimethoxy-(E/Z)-stilbene
IUPAC navn 1-methoxy-4-[2-(4-methoxyphenyl)vinyl]benzene
SMIL COc1ccc(C=Cc2ccc(OC)cc2)cc1
Tekniske data
Kemisk navn eller materiale 4,4'-Dimethoxystilbene
Mængde 100 mg
InChI formel InChI=1S/C16H16O2/c1-17-15-9-5-13(6-10-15)3-4-14-7-11-16(18-2)12-8-14/h3-12H,1-2H3
Formel vægt 240.115