missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxybenzhydrol, 98+%, Thermo Scientific Chemicals
283.19 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.
Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.
Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 69°C to 71°C |
Quantity | 10 g |
Beilstein | 748532 |
Solubility Information | Soluble in water (0.44g/L @ 25°C),methanol (0.1g/mL). |
Formula Weight | 244.29 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,4'-Dimethoxybenzhydrol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'